Imprimer
Sample storage convenience kits
Fournisseur: THERMO NATIONAL SCIENTIFIC
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
B7990-2 B7990-12A B7990-3 B7990-12 B7990-4
NSCAB7990-2EA 79.4 EUR
NSCAB7990-2 NSCAB7990-12A NSCAB7990-3 NSCAB7990-12 NSCAB7990-4
Sample storage convenience kits
Fioles
These sample storage unassembled convenience kits save time during sample preparation.
  • Solid top convenience kits include matched quantities of vials and screw caps with pre-assembled septa
  • Open top convenience kits contain shrink-wrapped vials and separately packaged caps and septa in polybags