Imprimer
Glass beads
Fournisseur: HILGENBERG
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
8413030-12 8413020-12 8413100-12
HILG8413030-12EA 364 EUR
HILG8413030-12 HILG8413020-12 HILG8413100-12
Glass beads
Perles Billes de verre
Glass beads made of soda lime glass.
  • Hydrologic resistance class 3
  • Acid resistance class 1
  • Alkali resistance class 2
Certifications: Class 3 (according to ISO 719), class 1 (according to DIN 12 116) and class 2 (according to ISO 695).