Imprimer
Cutter for glass tubing
Fournisseur: HILGENBERG
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
4018303 4018304
HILG4018303EA 139 EUR
HILG4018303 HILG4018304
Cutter for glass tubing
Cutters
This cutter is suitable for cutting and scoring glass tubing.
  • Ceramic blades and aluminium holder
  • Suitable to cut off the bottom of Mark-tubes