Imprimer
Cell culture media, CMRL 1066
Marques : Biowest
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
P0058-N1L P0058-N10L P0058-N5L
P0058-N1LEA 50.3 EUR
P0058-N1L P0058-N10L P0058-N5L
Cell culture media, CMRL 1066
Milieux de culture cellulaire
CRML was originally formulated for promoting strain L (Earle) cells under serum-free conditions. These media are also known for its ability to sustain cloning of monkey kidney cells and for growth of many other mammalian cell lines when supplemented with horse or calf serum.