Imprimer
Lymphocyte separation media, LymphoSep™
Fournisseur: Biowest
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
L0560-100 L0560-500
L0560-100EA 27.3 EUR
L0560-100 L0560-500
Lymphocyte separation media, LymphoSep™
Milieux de séparation cellulaire
Lymphosep is a standardised, high quality medium designed for simple and rapid isolation of PBMCs from treated whole blood. Lymphosep, like Ficoll™, has the optimised density 1,077 g/ml for lymphocyte separation to guarantee best and reliable results for your preparations.
High quality testing (bacteria in aerobic and anaerobic conditions, fungi and yeast, endotoxin) and instruction protocol available.
Certifications: CE marked