Imprimer
Filters for rotary vane pumps
Fournisseur: EDWARDS HIGH VACUUM METTLER
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
A46226000 A46229000 A46233000
531-0016EA 965 EUR
531-0016 181-0353 EDWAA46233000
Filters for rotary vane pumps
Filtres Filtres à gaz
Mist filters are used to separate and trap oil mist and dischage the odours with the exhaust from oil sealed rotary vacuum pumps.
  • Protection from oil mist emissions
  • Reduced oil level maintenance
  • Relief valve opens if oil element becomes blocked
  • Prevents pumps or pump oil from contaminaton
  • Efficient at 99,999% DOP test and are also azide proof
Filters are resistant to many chemicals, solvents and unaffected by alkalis, detergents and salt solutions. They are suitable for use with chemicals that form azide compounds.
Informations de livraison: Filters are supplied with NW25 clamp, centering ring and ‘O' ring and NW25 to (³/₄") BSP adaptor.