Imprimer
Re-caps, for blood collection tubes
Fournisseur: DELTALAB
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
308502 308602 308606
217-8901EA 98.3 EUR
217-8901 217-8903 217-8904
Re-caps, for blood collection tubes
Capuchons Bouchons pour tubes à essais
Ideal for re-capping blood collection tubes of vacuum type.
  • Manufactured in flexible polyethylene
  • Suitable for other glass and plastic tubes
  • Easy to cap and remove