Imprimer
Transport swabs
Fournisseur: DELTALAB
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
300295 300290 300291
DELT300295EA 75.9 EUR
DELT300295 DELT300290 DELT300291
Transport swabs
Écouvillons Écouvillons de transport
The modified Stuart medium allows the conservation and transport of a large number of pathological microorganisms
  • Fastidious organisms will remain viable for 24 hours and the others for several days
  • Medium reduced due to the presence of thioglycolate, which limits the enzymatic reactions of the bacteria
  • Multiplication of the bacteria prevented due to the lack of nitrogen in the media
Suitable for use with Neisseria gonorrhoeae, Heamophilus influenzae, Corynebacterium diphteriae, Trichomonas vaginalis, Streptococcus sp., Salmonella sp., Shigellas sp.
Cdt: In a peel-pack.
Attention: *Le(s) produit(s) sur cette page sont hors du champ d'application de la directive européenne 2019/904