Imprimer
Sampling spoons, eco-friendly, SP Bel-Art
Fournisseur: Bel-Art Products, a Part of SP
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
F369441000 F369421000 F369401000
BELAF369441000EA 91.2 EUR
BELAF369441000 BELAF369421000 BELAF369401000
Sampling spoons, eco-friendly, SP Bel-Art
Cuillères
PLA (polylactic acid) resin, eco-friendly, white.
  • Green alternative to petroleum-based sampling tools; helps meet sustainability goals
  • FDA grade spoons are safe for food, drug and cosmetic contact
  • Comparable in strength and durability to PS, PP and other plastic resin sampling tools
  • Long handle (17,8 cm) helps prevent personal contact with sampled material when reaching into bottles, jars and other confined areas
  • Rim of each bowl has a flat edge to assist in collecting and measuring small amounts with accuracy
The production of 100% plant-based PLA contributes less greenhouse gasses and uses less non renewable energy from source to resin than its petroleum-based counterparts. Disposal options include industrial composting for biodegradability, incineration, recycling and landfill. Use a separate spoon for each material sampled to avoid cross contamination.