Imprimer
Jars, wide mouth, with pressure cap
Fournisseur: LAMAPLAST
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
2177 2184 2182 2178 2175 2174 2180
LAMA2177EA 2.9 EUR
LAMA2177 LAMA2184 LAMA2182 LAMA2178 LAMA2175 LAMA2174 LAMA2180
Jars, wide mouth, with pressure cap
Pots Pots à usage général
Jars, made of PS, crystal clear, transparent body with LDPE pressure cap.
Wide mouth universal containers for creams, semi solid substances, crystals, powders and granulates.