Imprimer
Jerricans, narrow neck, HDPE, with screwed stopcock
Fournisseur: LAMAPLAST
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
3963 3961S
LAMA3963EA 63.3 EUR
LAMA3963 LAMA3961S
Jerricans, narrow neck, HDPE, with screwed stopcock
Flacons Jerricans
HDPE, natural.
  • Supplied with self sealing plastirob stopcock
  • Self sealing cap
  • Good chemical and temperature resistance
Ideal for use in food industries.