Imprimer
Digital block reactor, DRB 200
Fournisseur: Hach
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
LTV0829930001 LTV0829940001 LTV0829942001 LTV0829944001
HACHLTV0829930001EA 1460 EUR
HACHLTV0829930001 HACHLTV0829940001 HACHLTV0829942001 HACHLTV0829944001
Digital block reactor, DRB 200
Chauffe-blocs Bains à sec numériques
Digital block heaters which can hold one or two heating blocks, units allow cuvettes and reaction vessels to be digested at identical or different temperatures and time settings. Heating blocks enable cuvettes and reaction vessels to be digested at identical or different temperatures and time settings.
  • Pre-programmed for all standard digestions and freely programmable for user-specific digestions
  • Temperature stability
  • Excellent reproducibility
  • Simple handling, easy and safe to operate
  • Large, easily readable display and one-key operation for standard digestions