Imprimer
PCR tubes without caps, 0,2 ml, Axygen®
Fournisseur: Corning
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
PCR-02-NC PCR-02-NC-10K
732-0886EA 69.5 EUR
732-0886 AXYGPCR-02-NC-10K
PCR tubes without caps, 0,2 ml, Axygen®
Tubes Tubes PCR
0,2 ml polypropylene thin wall PCR tubes are designed to fit standard 96-well blocks. Ultra-thin and consistent wall thickness allows precise thermal transfer.
  • Certified free from DNase and RNase
  • Non pyrogenic
  • Non sterile