Imprimer
Bowls, with flat base and rim
Fournisseur: Adelphi Pharma Hygiene Products
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
BOW40.5P BOW4001P BOW4002P
COLDBOW40.5PEA 31.2 EUR
COLDBOW40.5P COLDBOW4001P COLDBOW4002P
Bowls, with flat base and rim
Plateaux/Récipients/Pots Plateaux et récipients de laboratoire
Stainless steel 304 grade.
  • Fast, easy cleaning
  • Good chemical resistance
  • Excellent thermal resistance
Bowls with flat base and rim are suitable for storage, mixing and other uses.