Imprimer
Precision hotplate, PZ
Fournisseur: GESTIGKEIT HARRY
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
PZ28-1 PZ28-2 PZ35 PZ60 PZ72
453-2131EA 2500 EUR
453-2131 453-2132 453-2133 453-2134 453-2135
Precision hotplate, PZ
Appareils chauffants Plaques chauffantes
Precision hotplates are equipped with adjustable temperature sensors +5 to +300 °C (PZ28-1 +30 to +110 °C). These sensors function as an emergency control which protect both the appliance and the substance to be heated against dangerous overheating.