Imprimer
Jars, water sampling, wide mouth, square
Fournisseur: DELTALAB
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
282340 282341 282350 282351
DELT282340EA 250 EUR
DELT282340 DELT282341 DELT282350 DELT282351
Jars, water sampling, wide mouth, square
Flacons Flacons d'échantillonnage de l'eau
PET, transparent, with red PP screw cap.
  • Available with or without thiosulphate sodium
  • Sterilised by radiation
Wide mouth and grips at two sides, easing the sample collection. Cap with internal liner made of polexan with a label that performs as tamper evident. Each bottle is labelled specifying description, code, lot, and expiry date.
With liquid sodium thiosulphate (24 mg/l): Ideal to analyse water meant for human consumption, swimming pools, and any other water where chlorine may modify the composition of the sample while being transported.
Empty, without thiosulphate: Ideal to sample water for physico-chemical analysis, as well as for other sampling requiring sterile bottles.