Imprimer
Stands for sedimentiation cones
Fournisseur: KOEHLER TECHNISCHE PRODUKTEN
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
A-1332 A-1331 A-1330
KOEHA-1332EA 546 EUR
KOEHA-1332 KOEHA-1331 KOEHA-1330
Stands for sedimentiation cones
Supports de statif
Racks, made of PP, for the storing of sedimentation vessels, stepless adjustable to height.
  • Thick-walled Chemical-resistant
  • Autoclavable up to 121 °C
  • Stand rods made of stainless high-grade steel
  • Even in filled condition, the rack can be safely transported with content