Imprimer
Culture tubes
Fournisseur: KLEINFELD
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
73500-18150 1217639
KLEI73500-18150EA 270 EUR
KLEI73500-18150 KLEI1217639
Culture tubes
Tubes Tubes de culture
Manufactured from 51 expansion borosilicate glass.
  • 51 expansion borosilicate glass offers excellent chemical resistance against sodium leaching, a factor common in soda lime glass products
  • Tubes are packed in convenient trays for ease of use on bench tops and in drawers
  • Trays are shrink-wrapped in plastic film, with tubes remaining in constant alignment for protection against in-transit breakage and contamination
  • The 6×50 mm size is often referred to as a 'Durham tube'
These premium quality tubes feature sturdy, uniform bottoms and consistent lengths. The 10×75 and 12×75 mm sizes are suitable for cell washing procedures.
Certifications: Glass conforming to USP Type I and ASTM E438, Type I, Class B requirements.