Imprimer
High vacuum angle valves, with sealed bellows, VA series
Fournisseur: Welch by Gardner Denver
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
700101 700371 700370 700109
ILMV700101EA 624 EUR
ILMV700101 ILMV700371 ILMV700370 ILMV700109
High vacuum angle valves, with sealed bellows, VA series
Vannes Vannes à vide
These high vacuum angle valves feature a very long life bellows-seal made from stainless steel.
  • High conductance
  • Leak rate 10-9 mbar L/s