Imprimer
Fine vacuum ball valves, manual
Fournisseur: Welch by Gardner Denver
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
700048 700047
ILMV700048EA 258 EUR
ILMV700048 ILMV700047
Fine vacuum ball valves, manual
Vannes Vannes à vide
These fine vacuum ball valves feature a small flange made of nickle plated aluminium.
  • Nickel-plated brass casing, polished brass ball, plastic (graphitised) seals