Imprimer
Overpressure relief valves, open
Fournisseur: Welch by Gardner Denver
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
700070 700069
ILMV700070EA 161 EUR
ILMV700070 ILMV700069
Overpressure relief valves, open
Vannes Soupapes de décompression
Small flange valve, open type, made of nickel-plated aluminium.
  • Prevents positive pressure rises over the maximum permissible level
  • Protects the pump against overload
  • Leak rate 10-2 mbar L/s
  • Opening pressure 1100 mbar (abs.) or 100 mbar (overpressure)
  • Polyamide valve body, Perbunan seals
  • Stainless steel compression springs
  • PTFE-coated aluminium sealing plate