Imprimer
Transport swabs, Transwab®
Fournisseur: MEDICAL WIRE EQUIPMENT
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
MW177S MW176S MW176SE2
MEDWMW177SEA 465 EUR
MEDWMW177S MEDWMW176S MEDWMW176SE2
Transport swabs, Transwab®
Écouvillons Écouvillons de transport
Transwab with a narrow shaft and bud for nasopharyngeal or urethral sampling.
  • Nasopharyngeal sampling such as for Bordetella pertussis
  • Open cell flow-through allows full access and release of sampled organisms
  • Optimum sample pick-up and release
Used in conventional and automated processing. Compatible with automated systems.
Attention: *Le(s) produit(s) sur cette page sont hors du champ d'application de la directive européenne 2019/904