This SD series dual stage rotary vane vacuum pump is designed and manufactured to meet the demanding requirements of various industrial and scientific applications. Suitable for several purposes (non-corrosive applications) such as manufacture of light bulbs, production of TV tubes, manufacture of electronic tubes, metallurgy, centrifuges, etc...
- Replacement shaft seal without dismantling
- Less air contamination and low oil consumption
- Alignment pins on pump module
- Easy maintenance and long lasting
Pump has a direct drive motor, making it very compact. An electrically insulated fold-away handle is used for easy carrying. The anit-suckback system ensures the tightness of the pump during accidental or voluntary shutdowns. The universal single-phase motor can be disassembled independently of the rest of the pump, without the need to drain the oil case. On the oil case, a vertical sight glass can be used to inspect the oil level easily when filling the tank and during the operation of the pump. A gas ballast enables the pumping of condensable vapours.
The inlet and exhaust end fittings are standardized. They are fitted vertically on the pump but can be positioned on the horizontal openings if required by operating conditions. The main replacement parts are interchangeable. This enables easier disassembly and assembly operations and replacement without changing the pumps performance. Various accessories can be used to adapt the pump to meet the requirements of your application.