Imprimer
Labels for petri dishes, PetriStickers™
Fournisseur: DIVERSIFIED BIOTECH
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
PSTK-1000 PSTK-1050 PSTK-1070 PSTK-1100 PSTK-2000 PSTK-2010 PSTK-2020 PSTK-2030
DIVEPSTK-1000EA 111 EUR
DIVEPSTK-1000 DIVEPSTK-1050 DIVEPSTK-1070 DIVEPSTK-1100 DIVEPSTK-2000 DIVEPSTK-2010 DIVEPSTK-2020 DIVEPSTK-2030
Labels for petri dishes, PetriStickers™
Étiquettes
PetriStickers™ allow for petri dishes to be labeled quickly, neatly and reliably.
  • Temperature resistant from +4 to +42° C
  • Water-resistant ink
Simply remove the label's protective backing and apply it to the back of a petri dish. After labeling, the dish can be held in a comfortable position for streaking or colony transfer, stored in an incubator or refrigerator, and photographed.