Imprimer
Screw caps for screw neck vials
Fournisseur: DELTALAB
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
918000 922000 928000
DELT918000EA 492 EUR
DELT918000 DELT922000 DELT928000
Screw caps for screw neck vials
Capuchons Bouchons pour flacons de chromatographie
These screw caps are made of autoclavable PP.
  • Internal silicone joint of pharmaceutical quality