Imprimer
GL-Aeration for bottles, BOLA
Fournisseur: Bohlender
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
N1697-14 N1697-18
BOHLN1697-14EA 47.1 EUR
BOHLN1697-14 BOHLN1697-18
GL-Aeration for bottles, BOLA
Filtres Filtres de ventilation
PPS screw cap and PTFE body.
  • Chemical resistant
  • Temperature resistant from –40 to +70 °C
Filter with PTFE membrane (pore size 0,2 µm) and flexible tubing, screw cap with GL threads. Application: For sterile pressure compensation on multiple distributors for bottles. Overpressure or vacuum in the vessel are prevented. A contamination of the product with dust or any other particles is avoided.
Attention: It is recommended to exchange the filter every six months.