Imprimer
Inoculating loops, Corning® Gosselin™
Fournisseur: Corning
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
OS10-02 OS1-01 OS1B-01 OS10B-02
612-3691EA 397 EUR
612-3691 612-3692 GOSSOS1B-01 GOSSOS10B-02
Inoculating loops, Corning® Gosselin™
Étaleurs Aiguilles et inoculateurs
These polystyrene inoculating loops are highly ergonomic and are color coded for easy identification.
  • Spherical or needle extremity
  • Assured sterile 10-3
Traceability information is provided on each bag.