Imprimer
High pressure fittings, for ¹/₃₂" tubing, Nanovolume™
Fournisseur: VICI JOUR RESEARCH
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
JR-ZGF1PK JR-ZGF.5PK JR-C-NEU.5XFPK JR-C-NEU.5FPK JR-C-NERU1FPK JR-C-EN.5FPKB JR-C-NNFFPK
JOURJR-ZGF1PKEA 0 EUR
JOURJR-ZGF1PK JOURJR-ZGF.5PK JOURJR-C-NEU.5XFPK JOURJR-C-NEU.5FPK JOURJR-C-NERU1FPK JOURJR-C-EN.5FPKB JOURJR-C-NNFFPK
High pressure fittings, for ¹/₃₂" tubing, Nanovolume™
Raccords et connecteurs
These fittings are designed for capillary LC, used in HTS techniques, proteomics, genomics, and other newer developments in industrial research requiring small sample size for reduction of analysis time, fittings and unions are easy to handle.
Nanovolume™ connectors include our one-piece fingertight fittings for ¹/₃₂" Ext.Ø tubing, featuring our patented collapsible ferrule. PEEK ferrules for use with our external nuts also feature the collapsible, grooved design.