Imprimer
Precolumn filters, high pressure
Fournisseur: VICI JOUR RESEARCH
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
JR-68262 JR-68258 JR-68264 JR-68264PE JR-1151-10P-5
JOURJR-68262EA 253 EUR
JOURJR-68262 JOURJR-68258 JOURJR-68264 JOURJR-68264PE JOURJR-1151-10P-5
Precolumn filters, high pressure
Filtres Filtres de précolonnes
Precolumn filter kit has a standard 10 to 32 fitting and fits direct to most column types without introducing an additional dead volume.
  • Minimum hold-up volume
  • Metal free PE frits and biocompatible titanium frits
It protects columns against fine particles, which may otherwise accumulate on the column frit, leading to split peaks and high backpressure.