Imprimer
Pipette tips, BagTips®
Fournisseur: INTERSCIENCE
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
252019 252024
432-0013EA 49.7 EUR
432-0013 INSC252024
Pipette tips, BagTips®
Pointes de pipettes
These sterile pipette tips are designed for use with the BagPipet® pipette. They are available in two sizes: Regular (19 cm) and Jumbo (24 cm).