Imprimer
Viscosity cups, Zahn
Fournisseur: Industrial Physics
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
VF2227 VF2230 VF2228 VF2226 VF2229
SIMEVF2227EA 270 EUR
SIMEVF2227 SIMEVF2230 SIMEVF2228 SIMEVF2226 SIMEVF2229
Viscosity cups, Zahn
Viscosimètres Coupes à écoulement
Stainless steel flow cup viscometers with fixed nozzle for measuring coatings and other fluids during application or production.
Certifications: ASTM D 1084, D 4212