Imprimer
Metric flangeless fittings, Upchurch Scientific®
Fournisseur: UPCHURCH SCIENTIFIC
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
P-362 P-363R P-207 P-207S P-226 P-247 P-288 P-301 P-307 P-307S P-307X P-326 P-342 P-342X P-343 P-343X P-347 P-347X P-353
UPCHP-362EA 15.3 EUR
UPCHP-362 UPCHP-363R UPCHP-207 UPCHP-207S UPCHP-226 UPCHP-247 UPCHP-288 UPCHP-301 UPCHP-307 UPCHP-307S UPCHP-307X UPCHP-326 UPCHP-342 UPCHP-342X UPCHP-343 UPCHP-343X UPCHP-347 UPCHP-347X UPCHP-353
Metric flangeless fittings, Upchurch Scientific®
Raccords et connecteurs
Metric flangeless fittings offer the same benefits as flangeless fittings with the additional feature of being used for metric tubing and ports. They are specifically designed to connect 1,8, 2,0, 2,5, 3,0, or 4,0 mm Ext.Ø tubing to flat-bottom ports when paired with the appropriate M6, 1/4-28, or 5/16-24 flangeless nuts.