Imprimer
Microplates, 96-well
Fournisseur: BOETTGER PAUL
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
05.011.0100 05.021.0100 05.031.0100
BOET05.011.0100EA 78.5 EUR
BOET05.011.0100 BOET05.021.0100 BOET05.031.0100
Microplates, 96-well
Microplaques Microplaques HTS
PS, optically clear, without lids. Microplates with a choice of different well types for different applications. The plates are fully compatible with standard reading and washing equipment.
  • Safe stacking, no danger of slipping
  • Gripping surfaces at the side of the plate
  • Frosted surface prevents errors from extraneous light sources
  • Alphanumerical well identification for easy sample location
  • Marking area on the side
  • Lids (optional) have condensation rings to reduce evaporation
  • Lid orientation corners reduce risk of cross-contamination