Imprimer
Ball valves, IBC
Fournisseur: BURKLE
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
0534-0001 0534-0002
BURK0534-0001EA 92.9 EUR
BURK0534-0001 BURK0534-0002
Ball valves, IBC
Robinets d'arrêt
The IBC discharge valve is a sturdy, high-quality ball valve with a hose connection and connection adapter including seal which is designed to fit all commonly used European IBC containers.
  • 1" flow
  • Maximum operating pressure 10 bar (at +20 °C)
  • Maximum operating temperature 80 °C
  • Fast and reliable residual emptying of the container with good dosing
Discharge valve and sealing cap PP fibreglass-reinforced, ball and adapter PP.
IBC discharge valve with 90° hose nozzle can be closed by the enclosed sealing cap suitable for flexible hoses.
IBC discharge valve with innovative hose screw connection attaches a wide range of hoses quickly, easily and reliably.