Imprimer
Safety cans
Fournisseur: BURKLE
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
2601-1001 2601-1002 2601-1005
BURK2601-1001EA 591 EUR
BURK2601-1001 BURK2601-1002 BURK2601-1005
Safety cans
Bidons de sécurité
Safety cans, made of high quality and chemically resistant stainless steel with PTFE gaskets.
  • Easy handling, ergonomic design and high stability
  • Flame traps and explosion-proof
  • Pressure relief valve (vent at about 0,3 bar)
  • Self-closing metering devices
Protection from exploding through flame protection and excess pressure valve, the containers have a high degree of chemical resistance. Can with removable flame protection in the filling connector and excess pressure valve in the self-closing fine dosage spout (pressure balance from about 0,3 bar).