Imprimer
Single channel pipettes, variable volume, Axygen® Axypet™
Fournisseur: Corning
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
AP-10 AP-100 AP-1000 AP-2 AP-20 AP-200 AP-50
AXYGAP-10EA 226 EUR
AXYGAP-10 AXYGAP-100 AXYGAP-1000 AXYGAP-2 AXYGAP-20 AXYGAP-200 AXYGAP-50
Single channel pipettes, variable volume, Axygen® Axypet™
Pipettes Pipetteurs
These high precision digital pipettes combine all the requirements of modern pipettes. Their body shape is equally comfortable in small and large hands, with design innovations such as two ejector buttons providing the user with the ability to optimise personal ergonomics. Constructed from durable PP/PVDF they are noticeably lighter in weight than many competitive models.
  • Thumb-friendly soft spring pipetting system reduces fatigue
  • Unique EZIJECT side ejection option reduces thumb movement
  • Finger-hook to take the weight, for a more relaxed grip
  • Large ergonomic push button is easy on the thumb during contact
  • Tip ejector incorporates height adjusters to accommodate virtually all brands of tips
  • Fully autoclavable