Imprimer
Chlorure de benzylhexadécyldiméthylammonium 95%
Marques : Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Danger
Synonyme(s): Cetalkonium chloride, N-Benzyl-N,N-dimethylhexadecan-1-aminium chloride
32377.18 32377.30
32377.18EA 141 EUR
32377.18 32377.30
Chlorure de benzylhexadécyldiméthylammonium 95%
Chlorure de benzylhexadécyldiméthylammonium
Benzyldimethylhexadecylammonium chloride has been used in a study to assess the removal of cationic surfactants from aqueous solutions using activated carbon cloth (ACC). It has also been used in a study to investigate the prototropic behavior of the dye, 7-hydroxy-4-methylcoumarin (7H4MC).
Purity: 95% CAS No.: 122-18-9 Molecular Formula: C25H46ClN.xH2O Formula Weight: 396.10(anhy) Physical Form: Powder Appearance: White Melting Point: 54-58° Beilstein Reference: 3802341 Merck Reference: 14,2017 UN No.: 3261 MDL No.: MFCD00011625
Formule: C₂₅H₄₆ClN
Poids moléculaire: 396.10 g/mol
Point de fusion: 54…58 °C
Température de stockage: Température ambiante
Numéro MDL: MFCD00011625
Numéro CAS: 122-18-9
EINECS: 204-526-3
UN: 3261
ADR: 8 III
Indice Merck: 14,2017