Imprimer

Pilocarpine hydrochloride, poudre blanche à écoulement libre

Marques : MP Biomedicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

Danger

Synonyme(s): (+)-Pilocarpine hydrochloride

0215189210 0215189201 0215189205
ICNA0215189210EA 326 EUR
ICNA0215189210 ICNA0215189201 ICNA0215189205
Pilocarpine hydrochloride, poudre blanche à écoulement libre
Pilocarpine hydrochloride

Pilocarpine hydrochloride is used in neuroscience and opthamology research. It is a parasympathomimetic agent with activity similar to that of acetylcholine.


Soluble in water (100 mg/ml) with heat as needed, yielding a clear, colourless solution. It is also soluble in alcohol and slightly soluble in chloroform.


Attention: Research Use Only (RUO).

Formule: C₁₁H₁₇ClN₂O₂
Température de stockage: Température ambiante
Numéro MDL: MFCD00012722
Numéro CAS: 54-71-7
EINECS: 200-212-5

Commander maintenant

Résultats des tests de spécification

Identity Test Passes
FTIR Conforms to Standard
Melting Point 196 - 206°C
Moisture Content ≤3,0%
Optical Rotation +86° to +96° (c=2, water)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR