Imprimer
Beakers, low form, LDPE
Fournisseur: DELTALAB
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
242802 242803 242805 242806 242811
DELT242802EA 554 EUR
DELT242802 DELT242803 DELT242805 DELT242806 DELT242811
Beakers, low form, LDPE
Béchers
LDPE, translucent.
  • Good chemical resistance
  • Good temperature resistance
  • 1000 ml beaker comes with lid in one bag
For use with liquid and solid samples.