Imprimer
Transport swab with Cary Blair medium
Fournisseur: DELTALAB
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
300280 300280.2
DELT300280EA 103 EUR
DELT300280 DELT300280.2
Transport swab with Cary Blair medium
Écouvillons Écouvillons de transport
The glycerophosphate has been substituted by inorganic phosphate since glycerophosphate is a metabolite for some bacteria, so they could grow and disgrise the pathogens amount.
  • Sterilised by radiation
  • Methylene blue removed with pH increased to 8,4
  • Cary Blair media for the conservation of pathogens including Neisseria sp., Haemophilus sp. or Streptococcus sp.
Originally developed for faecal samples to transport anaerobic bacteria.
Attention: *Le(s) produit(s) sur cette page sont hors du champ d'application de la directive européenne 2019/904