Imprimer
Dialysis tubing, VISKING®
Fournisseur: Serva
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
44104.01 44104.02 44104.04 44110.01 44110.02 44110.04 44114.01 44114.02 44114.04 44120.01 44120.02 44120.05 44126.02 44126.03 44130.01 44130.02 44130.03
SERA44104.01EA 102 EUR
SERA44104.01 518-1120 SERA44104.04 SERA44110.01 SERA44110.02 SERA44110.04 SERA44114.01 SERA44114.02 SERA44114.04 SERA44120.01 SERA44120.02 SERA44120.05 SERA44126.02 SERA44126.03 SERA44130.01 SERA44130.02 SERA44130.03
Dialysis tubing, VISKING®
Tuyaux Membranes de dialyse
MWCO 12 to 14 kD, regenerated cellulose.
  • Resistant to chemicals
  • Contain low level of heavy metal and sulfide impurities
  • Suitable for pH range 2 to 12 and temperatures 4 to 60 °C