Imprimer
Drigalski spreaders, L-shaped with curved end
Fournisseur: DELTALAB
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
200500.1 200510.1
391-3018EA 246 EUR
391-3018 391-3019
Drigalski spreaders, L-shaped with curved end
Étaleurs Étaleurs de cellules
Designed for surface spreading. Made of white PS. Sterilised by ethylene oxide.
  • Base length allows spreading liquid samples over any type of Petri dish and avoids contact with the dish walls
  • Curved spreader extremity and rounded corners avoid damaging media surface while spreading
  • Bent and rounded edges minimize the possibility of breakage of the solid medium during extension
Can be used in places where sterilisation before use is not possible (for instance when samples are collected far from the lab).
Cdt: Available in peel-pack containing 1 or 5 units, marked with code, expiry date, lot and sterilisation method.