Imprimer
Tissue grinders
Fournisseur: DELTALAB
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
196102 196105 196110 196115 196302 196305 196310 196315
432-1202EA 138 EUR
432-1202 432-1205 432-1210 432-1215 432-1235 432-1240 432-1245 432-1250
Tissue grinders
Homogénéisateurs Broyeurs pour tissus
Clearance between Glass vessel and appropriately sized plunger is approximately 200 µm.