Imprimer
Staining baskets and racks
Fournisseur: DELTALAB
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
CP-30H CP-30 CP-45H
631-9740EA 129 EUR
631-9740 DELTCP-30 DELTCP-45H
Staining baskets and racks
Portoirs Portoirs à coloration
Staining baskets and racks, made of stainless steel. Designed for 76×26 mm slides.
  • Good chemical resistance
  • Good temperature resistance