Imprimer
Balloons, with Hose Connection
Fournisseur: DEUTSCH NEUMANN
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
4500002 4500003
DENE4500002EA 178 EUR
DENE4500002 201-0907
Balloons, with Hose Connection
Bags Gas Bags
These are red natural rubber latex balloon suitable for rotary evaporators for mixing or collecting gas and for controlling air or gas flows at suitable equipment. It also features a tubing with length 140 mm and diameter 12 mm.
Hardness (Shore A): 40