Imprimer
Capillary columns, with SafeGuard, TraceGOLD™ TG-WaxMS
Fournisseur: Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
26088-1421 26088-1435
555-8589EA 997 EUR
555-8589 555-8587
Capillary columns, with SafeGuard, TraceGOLD™ TG-WaxMS
Colonnes pour chromatographie
These columns are manufactured for better column-to-column reproducibility.
  • Provide prolong lifetime for column
  • Polar phase, polyethylene glycol
  • High thermal stability - maximum temperatures up to 260 °C
  • Resists oxidative damage, damage from strongly acidic or basic volatiles better than silicone solid phases