Imprimer
Gas and vapour samplers
Fournisseur: Trajan Scientific and Medical
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
090111 090112
SGEA090111EA 772 EUR
SGEA090111 SGEA090112
Gas and vapour samplers
Samplers Gas Samplers
This gas and vapour sampler enables spot collection of environmental gas samples or low pressure gas streams in 200 cm³ impervious resealable bags.
Informations de commande: Includes a canister, a bulb, 10 bags, a marker pen and instructions.