Imprimer
Hydrophobic interaction chromatography columns, ProPac™ HIC-10
Fournisseur: Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
074197 063653 063655 063665
555-7490EA 1362.56 EUR
555-7490 555-7295 555-7296 555-7297
Hydrophobic interaction chromatography columns, ProPac™ HIC-10
Colonnes pour chromatographie
These columns are used for the high resolution separation of the proteins and peptides.
  • Proteins are separated under non-denaturing conditions
  • High protein loading capacity for protein purification applications