Imprimer
Drums, wide neck, nestable, with screw lid, UN approved
Fournisseur: CURTEC
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
7230-98-855 7240-98-855 7250-98-855 7260-98-855 7276-98-855
216-1366EA 67.5 EUR
216-1366 216-1367 216-1368 216-1369 CURT7276-98-855
Drums, wide neck, nestable, with screw lid, UN approved
Barrels, Kegs, Drums
Drums, HDPE, nestable, white with blue screw lid.
  • Water tight
  • Tamper evident
  • Stackable
  • UN marking (1H2) for hazardous solids
  • Food grade
For storage and shipment of powders, pastes and liquids. Take up less storage space when empty and help reduce your logistic cost.