Imprimer
Capillary columns, with SafeGuard, TraceGOLD™ TG-5MS
Fournisseur: Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
26098-0475 26098-1305 26098-1425 26098-1435 26098-5785
555-8691EA 1070 EUR
555-8691 555-8689 555-8685 555-8683 555-9336
Capillary columns, with SafeGuard, TraceGOLD™ TG-5MS
Colonnes pour chromatographie
Exceptionally low bleed for optimal signal-to-noise ratio, sensitivity and MS integrity.
  • Provide prolong lifetime for column
  • Low polarity phase, 5% diphenyl/95% dimethylm polysiloxane
  • High thermal stability - maximum temperatures up to 330/350 °C
  • Ideal for analysis of active compounds